Assay Details
| Gene Symbol: Trappc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021789.2 | NP_068561.1 | 4 - 5 | 831 | 89 | |
| GenBank mRNA | AF233340.1 | - | 4 - 5 | 788 | 89 | |
| AK032168.1 | - | 4 - 5 | 597 | 89 | ||
| BC038898.1 | - | 4 - 5 | 831 | 5401311 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467467 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467466 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467464 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467465 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | AF233340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | AK005276.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | AK032168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | BC038898.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | NM_021789.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | AF233340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | AK005276.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | AK032168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | BC038898.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407904 | NM_021789.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | AJ000262.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | AJ002272.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | AK138436.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | AK196068.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | BC034089.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | BC053043.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635105 | NM_010404.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35512 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132696 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | AF233340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | AK005276.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | AK032168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | BC038898.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | NM_021789.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | AF233340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | AK005276.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | AK032168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | BC038898.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407904 | NM_021789.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467444 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132696 | - | - |
| Whole Mouse Genome G4122A | A_52_P641172 | AF233340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641172 | AK005276.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641172 | AK032168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641172 | BC038898.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641172 | NM_021789.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | AF233340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | AK005276.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | AK032168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | BC038898.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | NM_021789.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | AF233340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | AK005276.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | AK032168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | BC038898.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407904 | NM_021789.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111181 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07518233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093114 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467464 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467467 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467465 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08061282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07639134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07710221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467451 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592855 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429632_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415674_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592850 | - | - |
| Mouse Genome 430 2.0 Array | 1429632_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415674_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415674_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429632_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 60409_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 60409_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900017975.mm.1 | NM_021789.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017975.mm.1 | NM_021789.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017975.mm.1 | TC0900002164.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415674_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415674_a_at | - | - |
| Mouse Expression Set 430 | 1415674_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415674_a_at | - | - |
| Murine Genome U74v2 Set | 160361_PM_at | - | - |
| Murine Genome U74v2 Set | 160361_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415674_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415674_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415674_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415674_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429632_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429632_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415674_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415674_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 785123 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 520044 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 520044 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225507 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225507 | - | - |
| mouse-6 v1.1 (Illumina) | scl35956.2.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520044 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35956.2.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520044 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2790573 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225507 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2790573 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225507 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl35956.2.1_28-S | - | - |

Trappc4
Genomic Map