Assay Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023138.4 | NP_075627.2 | 4 - 5 | 730 | 79 | |
| GenBank mRNA | AK009392.1 | - | 4 - 5 | 747 | 79 | |
| AK080574.1 | - | 4 - 5 | 731 | 79 | ||
| AK143425.1 | - | 4 - 5 | 710 | 79 | ||
| AK171584.1 | - | 4 - 5 | 722 | 79 | ||
| AK172055.1 | - | 4 - 5 | 731 | 79 | ||
| AK181608.1 | - | 2 - 3 | 182 | 79 | ||
| AK203356.1 | - | 2 - 3 | 182 | 79 | ||
| BC014830.1 | - | 4 - 5 | 712 | 4483473 | 79 | |
| S68267.1 | - | 4 - 5 | 651 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k2
Genomic Map