Assay Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023138.4 | NP_075627.2 | 7 - 8 | 1121 | 84 | |
| GenBank mRNA | AK009392.1 | - | 7 - 8 | 1138 | 84 | |
| AK080574.1 | - | 7 - 8 | 1122 | 84 | ||
| S68267.1 | - | 7 - 8 | 1042 | 84 | ||
More Information
Set Membership: |
|

Map2k2
Genomic Map