Assay Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164222.1 | NP_001157694.1 | 5 - 6 | 1021 | 72 | |
| NM_023420.2 | NP_075909.1 | 5 - 6 | 1021 | 72 | ||
| GenBank mRNA | AF232932.1 | - | 5 - 6 | 1035 | 72 | |
| AF232934.1 | - | 5 - 6 | 1035 | 72 | ||
| AK012989.1 | - | 5 - 6 | 1002 | 72 | ||
| AK030305.1 | - | 6 - 7 | 929 | 72 | ||
| AK038061.1 | - | 5 - 6 | 1011 | 72 | ||
| AK038803.1 | - | 5 - 6 | 1002 | 72 | ||
| AK047100.1 | - | 5 - 6 | 1018 | 72 | ||
| AK080958.1 | - | 5 - 6 | 1019 | 72 | ||
| AK163624.1 | - | 5 - 6 | 1011 | 72 | ||
| BC016197.1 | - | 5 - 6 | 984 | 4502118 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Col4a3bp
Genomic Map