Assay Details
| Gene Symbol: Tnfrsf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023680.3 | NP_076169.2 | 6 - 7 | 570 | 84 | |
| GenBank mRNA | AJ278265.1 | - | 6 - 7 | 84 | ||
| AK012838.1 | - | 6 - 7 | 571 | 84 | ||
| AY165626.1 | - | 6 - 7 | 593 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155197 | AY046551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155197 | AY165626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155197 | AY165627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155197 | BC076592.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155197 | NM_023680.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | AY046551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | BC076592.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | NM_023680.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | AK012838.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | AY046551.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | BC076592.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | NM_023680.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323852 | AK012838.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68650 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112663 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | AY046551.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | BC076592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | NM_023680.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | AK012838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | AY046551.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | BC076592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | NM_023680.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323852 | AK012838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155197 | AY046551.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155197 | AY165626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155197 | AY165627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155197 | BC076592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155197 | NM_023680.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25602226 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06402282 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01659619 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112663 | - | - |
| Whole Mouse Genome G4122A | A_52_P323852 | AY046551.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | BC076592.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | NM_023680.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | AK012838.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | AY046551.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | BC076592.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | NM_023680.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323852 | AK012838.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134272 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135001 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06436300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25602178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06436301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06437525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25602178 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10569503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10569496 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422039_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422038_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426095_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10569494 | - | - |
| Mouse Genome 430 2.0 Array | 1426095_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426095_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422039_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422038_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422038_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422039_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 79202_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 79202_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700038819.mm.1 | NM_023680.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038819.mm.1 | TC0700004592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700038820.mm.1 | NM_023680.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038820.mm.1 | TC0700004592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700038822.mm.1 | NM_023680.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038822.mm.1 | TC0700004592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700038824.mm.1 | NM_023680.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038824.mm.1 | TC0700004592.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700038823.mm.1 | NM_023680.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038823.mm.1 | TC0700004592.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422039_at | - | - |
| Mouse Expression Set 430 | 1422038_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426095_a_at | - | - |
| Mouse Expression Set 430 | 1422038_a_at | - | - |
| Mouse Expression Set 430 | 1422039_PM_at | - | - |
| Mouse Expression Set 430 | 1426095_PM_a_at | - | - |
| Mouse Expression Set 430 | 1442590_PM_at | - | - |
| Mouse Expression Set 430 | 1442590_at | - | - |
| Murine Genome U74v2 Set | 129236_at | - | - |
| Murine Genome U74v2 Set | 129236_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426095_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426095_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422039_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422038_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422038_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422039_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422039_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422038_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426095_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426095_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422039_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422038_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5890731 | - | - |
| MouseWG-6 v2 Expression BeadChip | 105570722 | - | - |
| MouseWG-6 v2 Expression BeadChip | 770400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105570722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5890731 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5050050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005050050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005890731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219749 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2454204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2467549 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2454204 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2467549 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219749 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2522818 | - | - |
| mouse-6 v1.1 (Illumina) | scl0319679.1_12-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0079202.2_327-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00319679.1_98-S | - | - |
| mouse-6 v1.1 (Illumina) | scl079202.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5050050 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770400 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5890731 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105570722 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0079202.2_327-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319679.1_12-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl079202.1_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050050 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770400 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105570722 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2467549 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2892413 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2454204 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2522818 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2467549 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2454204 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2892413 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0079202.2_327-S | - | - |

Tnfrsf22
Genomic Map