Assay Details
| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286005.1 | NP_001272934.1 | 7 - 8 | 1063 | 63 | |
| NM_026180.3 | NP_080456.1 | 8 - 9 | 1315 | 63 | ||
| NR_104382.1 | 8 - 9 | 1312 | 63 | |||
| GenBank mRNA | AF324495.1 | - | 8 - 9 | 1305 | 63 | |
| AK004871.1 | - | 8 - 9 | 1272 | 63 | ||
| AK050938.1 | - | 7 - 8 | 1065 | 63 | ||
| AY196215.1 | - | 8 - 9 | 1302 | 63 | ||
| AY196216.1 | - | 8 - 9 | 1302 | 63 | ||
| BC138484.1 | - | 8 - 9 | 1279 | 8861504 | 63 | |
| BC145293.1 | - | 8 - 9 | 1279 | 9053847 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg8
Genomic Map