Assay Details
| Gene Symbol: Snx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026386.1 | NP_080662.1 | 5 - 6 | 522 | 122 | |
| GenBank mRNA | AK002692.1 | - | 5 - 6 | 522 | 122 | |
| AK005470.1 | - | 5 - 6 | 534 | 122 | ||
| AK010572.1 | - | 5 - 6 | 519 | 122 | ||
| AK075929.1 | - | 5 - 6 | 519 | 122 | ||
| AK145840.1 | - | 5 - 6 | 573 | 122 | ||
| BC006960.1 | - | 5 - 6 | 522 | 2812557 | 122 | |
| CT010229.1 | - | 5 - 6 | 507 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snx2
Genomic Map