Assay Details
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004878.1 | - | 1 - 2 | 90 | 85 | |
AK159405.1 | - | 1 - 2 | 79 | 85 | ||
BB872569.1 | - | 1 - 2 | 131 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK004878.1, AK159405.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
