Assay Details
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030239.2 | NP_084515.2 | 11 - 12 | 1527 | 125 | |
GenBank mRNA | AF324242.1 | - | 11 - 12 | 1479 | 125 | |
AK030831.1 | - | 11 - 12 | 1529 | 125 | ||
AK051880.1 | - | 11 - 12 | 1493 | 125 | ||
AK156626.1 | - | 11 - 12 | 1495 | 125 | ||
BC138772.1 | - | 11 - 12 | 1508 | 8861794 | 125 | |
BC138773.1 | - | 11 - 12 | 1507 | 8861795 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
