Assay Details
| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030709.2 | NP_109634.2 | 10 - 11 | 1144 | 77 | |
| GenBank mRNA | AB016229.1 | - | 10 - 11 | 1142 | 77 | |
| AB016230.1 | - | 10 - 11 | 1144 | 77 | ||
| AB016423.1 | - | 6 - 7 | 759 | 77 | ||
| AB041037.1 | - | 10 - 11 | 1101 | 77 | ||
| AK029714.1 | - | 11 - 12 | 1991 | 77 | ||
| BC132151.1 | - | 9 - 10 | 1090 | 40130428 | 77 | |
| BC145027.1 | - | 9 - 10 | 1072 | 9053575 | 77 | |
More Information
Set Membership: |
|

Tmprss5
Genomic Map