Assay Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030721.2 | NP_109646.2 | 4 - 5 | 566 | 71 | |
| GenBank mRNA | AJ278430.1 | - | 4 - 5 | 599 | 71 | |
| AK028818.1 | - | 4 - 5 | 987 | 71 | ||
| AK053960.1 | - | 4 - 5 | 563 | 71 | ||
| AK128922.1 | - | 4 - 5 | 526 | 71 | ||
| AK154112.1 | - | 3 - 4 | 657 | 71 | ||
| AK171598.1 | - | 549 | 71 | |||
| AK194193.1 | - | 1 - 2 | 204 | 71 | ||
| BC044725.2 | - | 4 - 5 | 586 | 5374281 | 71 | |
| BC055019.1 | - | 4 - 5 | 566 | 4162223 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox3
Genomic Map