Assay Details
| Gene Symbol: Il6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031168.1 | NP_112445.1 | 3 - 4 | 347 | 124 | |
| GenBank mRNA | AK089780.1 | - | 3 - 4 | 366 | 124 | |
| AK150440.1 | - | 3 - 4 | 366 | 124 | ||
| AK152189.1 | - | 2 - 3 | 353 | 124 | ||
| BC132458.1 | - | 3 - 4 | 340 | 40130735 | 124 | |
| BC138766.1 | - | 3 - 4 | 340 | 8861788 | 124 | |
| BC145409.1 | - | 3 - 4 | 340 | 124 | ||
| DQ788722.1 | - | 3 - 4 | 316 | 124 | ||
| J03783.1 | - | 3 - 4 | 364 | 124 | ||
| X06203.1 | - | 3 - 4 | 349 | 124 | ||
| X54542.1 | - | 3 - 4 | 347 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il6
Genomic Map