Assay Details
| Gene Symbol: Myog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031189.2 | NP_112466.1 | 1 - 2 | 522 | 69 | |
| GenBank mRNA | AB221643.1 | - | 1 - 2 | 470 | 69 | |
| BC048683.1 | - | 1 - 2 | 537 | 6516364 | 69 | |
| BC068019.1 | - | 1 - 2 | 503 | 6508229 | 69 | |
| D90156.1 | - | 1 - 2 | 513 | 69 | ||
| X15784.1 | - | 1 - 2 | 68 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Wnt10b deficiency promotes coexpression of myogenic and adipogenic programs in myoblasts.
- Authors
- Vertino AM, Taylor-Jones JM, Longo KA, Bearden ED, Lane TF, McGehee RE Jr, MacDougald OA, Peterson CA
- Journal
- Mol Biol Cell - Page No 2039 (Fri Apr 01 00:00:00 EST 2005)
- Volume (Year)
- 16 (2005)
- Product Usage
- -

Myog
Genomic Map