Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173372.1 | NP_001166843.1 | 1 - 2 | 44 | 69 | |
| NM_033648.1 | NP_387468.1 | 1 - 2 | 44 | 69 | ||
| GenBank mRNA | AK018728.1 | - | 1 - 2 | 44 | 69 | |
| AK054004.1 | - | 1 - 2 | 45 | 69 | ||
| BC086918.1 | - | 2 - 3 | 305 | 6762406 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018728.1, BC086918.1, NM_001173372.1, NM_033648.1 transcripts.
Set Membership: |
|

Fxyd4
Genomic Map