| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600022991.mm.1 |
TC0600002992.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022991.mm.1 |
NM_001173372.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022991.mm.1 |
NM_001173372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022991.mm.1 |
NM_033648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022991.mm.1 |
NM_033648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022994.mm.1 |
TC0600002992.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022994.mm.1 |
NM_001173372.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022994.mm.1 |
NM_001173372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022994.mm.1 |
NM_033648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022994.mm.1 |
NM_033648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022995.mm.1 |
TC0600002992.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022995.mm.1 |
NM_001173372.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022995.mm.1 |
NM_001173372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022995.mm.1 |
NM_033648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022995.mm.1 |
NM_033648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022997.mm.1 |
TC0600002992.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022996.mm.1 |
TC0600002992.mm.1 |
MTA TC |