Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173372.1 | NP_001166843.1 | 4 - 5 | 184 | 70 | |
| NM_033648.1 | NP_387468.1 | 4 - 5 | 187 | 70 | ||
| GenBank mRNA | AK018728.1 | - | 4 - 5 | 184 | 70 | |
| BC086918.1 | - | 5 - 6 | 448 | 6762406 | 70 | |
More Information
Set Membership: |
|

Fxyd4
Genomic Map