Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173372.1 | NP_001166843.1 | 6 - 7 | 286 | 61 | |
| NM_033648.1 | NP_387468.1 | 6 - 7 | 289 | 61 | ||
| GenBank mRNA | AK018728.1 | - | 6 - 7 | 286 | 61 | |
| AU041511.1 | - | 3 - 4 | 61 | |||
| BC086918.1 | - | 7 - 8 | 550 | 6762406 | 61 | |
More Information
Set Membership: |
|

Fxyd4
Genomic Map