Assay Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053198.3 | NP_444428.3 | 13 - 14 | 961 | 144 | |
| GenBank mRNA | AF325263.1 | - | 13 - 14 | 903 | 144 | |
| AK148530.1 | - | 12 - 13 | 930 | 144 | ||
| BC058211.1 | - | 14 - 15 | 1153 | 144 | ||
| BC089535.1 | - | 13 - 14 | 943 | 6589805 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35875 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155292 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117496 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32227605 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09220796 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09220810 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09220837 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117496 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155292 | - | - |
| Whole Mouse Genome G4122A | A_52_P599680 | - | - |
| Whole Mouse Genome G4122A | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320980 | AF325263.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111496 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117496 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14171227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14147357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14147361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32227540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14300069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14300070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14200847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14157087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32227587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14171227 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468857 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468858 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468859 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468860 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468854 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418873_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468853 | - | - |
| Mouse Genome 430 2.0 Array | 1418873_at | - | - |
| Mouse Genome 430 2.0 Array | 1418873_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94281_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94281_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900015476.mm.1 | NM_053198.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015476.mm.1 | NM_053198.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015476.mm.1 | TC1900001753.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900015479.mm.1 | NM_053198.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015479.mm.1 | NM_053198.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015479.mm.1 | TC1900001753.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418873_PM_at | - | - |
| Mouse Expression Set 430 | 1418873_at | - | - |
| Murine Genome U74v2 Set | 167294_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167294_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418873_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418873_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418873_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418873_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6420176 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4560059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6420176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560059 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248840 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776377 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248840 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776377 | - | - |
| mouse-6 v1.1 (Illumina) | scl094281.1_62-S | - | - |
| mouse-6 v1.1 (Illumina) | scl52316.14.1_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420176 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560059 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl094281.1_62-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52316.14.1_45-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420176 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560059 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2776377 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248840 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248840 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776377 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl094281.1_62-S | - | - |

Sfxn4
Genomic Map