Assay Details
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080639.3 | NP_542370.3 | 3 - 4 | 421 | 83 | |
GenBank mRNA | AF282730.1 | - | 3 - 4 | 352 | 83 | |
AK038820.1 | - | 3 - 4 | 420 | 83 | ||
AK040814.1 | - | 3 - 4 | 420 | 83 | ||
AK079347.1 | - | 3 - 4 | 422 | 83 | ||
AK140367.1 | - | 3 - 4 | 421 | 83 | ||
BC064046.1 | - | 3 - 4 | 401 | 875422 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
