Assay Details
| Gene Symbol: Slc6a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133661.3 | NP_598422.2 | 2 - 3 | 529 | 78 | |
| GenBank mRNA | BC019211.1 | - | 2 - 3 | 533 | 5053116 | 78 |
| M97632.1 | - | 2 - 3 | 269 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a12
Genomic Map