Assay Details
Gene Symbol: Trf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133977.2 | NP_598738.1 | 10 - 11 | 1356 | 97 | |
GenBank mRNA | AF440692.1 | - | 9 - 10 | 1286 | 97 | |
AK085754.1 | - | 10 - 11 | 1341 | 97 | ||
AK142599.1 | - | 1346 | 97 | |||
AK146549.1 | - | 10 - 11 | 1341 | 97 | ||
AK149559.1 | - | 10 - 11 | 1341 | 97 | ||
AK149595.1 | - | 10 - 11 | 1341 | 97 | ||
AK150782.1 | - | 10 - 11 | 1389 | 97 | ||
AK168368.1 | - | 10 - 11 | 1343 | 97 | ||
AK168405.1 | - | 10 - 11 | 1343 | 97 | ||
AK168419.1 | - | 10 - 11 | 1341 | 97 | ||
AK168938.1 | - | 10 - 11 | 1341 | 97 | ||
AK169187.1 | - | 10 - 11 | 1340 | 97 | ||
BC008559.1 | - | 8 - 9 | 1078 | 97 | ||
BC012313.1 | - | 10 - 11 | 1356 | 3981648 | 97 | |
BC018573.1 | - | 5 - 6 | 626 | 97 | ||
BC020295.1 | - | 10 - 11 | 1325 | 97 | ||
BC022986.1 | - | 10 - 11 | 1334 | 5123741 | 97 | |
BC058216.1 | - | 10 - 11 | 1322 | 97 | ||
BC058218.1 | - | 10 - 11 | 1335 | 97 | ||
BC092046.1 | - | 10 - 11 | 1317 | 3594705 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
