Assay Details
| Gene Symbol: Il13ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080147.1 | - | 8 - 9 | 1034 | 71 | |
| AK089984.1 | - | 8 - 9 | 1065 | 71 | ||
| AK143292.1 | - | 8 - 9 | 71 | |||
| AK154675.1 | - | 8 - 9 | 1090 | 71 | ||
| BC021472.1 | - | 4 - 5 | 488 | 71 | ||
| BC052425.1 | - | 8 - 9 | 1057 | 6835500 | 71 | |
| BC059939.1 | - | 8 - 9 | 1025 | 6477224 | 71 | |
| S80963.1 | - | 8 - 9 | 1062 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il13ra1
Genomic Map