Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200004331.mm.1 |
NM_138748.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004331.mm.1 |
NM_138748.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004331.mm.1 |
TC0200000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004332.mm.1 |
NM_138748.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004332.mm.1 |
NM_138748.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004332.mm.1 |
TC0200000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004333.mm.1 |
NM_138748.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004333.mm.1 |
NM_138748.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004333.mm.1 |
TC0200000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004334.mm.1 |
NM_138748.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004334.mm.1 |
NM_138748.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004334.mm.1 |
TC0200000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004326.mm.1 |
NM_138748.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004326.mm.1 |
NM_138748.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004326.mm.1 |
TC0200000535.mm.1 |
MTA TC |