Assay Details
| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138952.3 | NP_620402.1 | 2 - 3 | 427 | 116 | |
| GenBank mRNA | AF461040.1 | - | 2 - 3 | 325 | 116 | |
| AF487539.1 | - | 2 - 3 | 325 | 116 | ||
| AK159593.1 | - | 2 - 3 | 451 | 116 | ||
| AK172319.1 | - | 2 - 3 | 449 | 116 | ||
| AK172417.1 | - | 2 - 3 | 449 | 116 | ||
| BC050845.1 | - | 2 - 3 | 440 | 116 | ||
| BC058088.1 | - | 2 - 3 | 572 | 116 | ||
| BC069878.1 | - | 2 - 3 | 452 | 5316066 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ripk2
Genomic Map