Assay Details
| Gene Symbol: Slc6a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144512.2 | NP_653095.1 | 11 - 12 | 1355 | 61 | |
| GenBank mRNA | AK149557.1 | - | 11 - 12 | 1363 | 61 | |
| BC021606.1 | - | 2 - 3 | 213 | 61 | ||
| BC023117.1 | - | 11 - 12 | 1343 | 4240641 | 61 | |
| BC029637.1 | - | 11 - 12 | 1355 | 4195373 | 61 | |
More Information
Set Membership: |
|

Slc6a13
Genomic Map