Assay Details
| Gene Symbol: Sec14l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144520.2 | NP_653103.1 | 5 - 6 | 468 | 87 | |
| GenBank mRNA | BC005759.1 | - | 5 - 6 | 468 | 2655022 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l2
Genomic Map