Assay Details
| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010348.3 | NP_034478.1 | 4 - 5 | 1286 | 83 | |
| NM_146072.4 | NP_666184.2 | 4 - 5 | 1286 | 83 | ||
| GenBank mRNA | AK042635.1 | - | 4 - 5 | 1287 | 83 | |
| AK043542.1 | - | 4 - 5 | 1266 | 83 | ||
| AK043737.1 | - | 4 - 5 | 1250 | 83 | ||
| AK044250.1 | - | 4 - 5 | 1245 | 83 | ||
| AK044420.1 | - | 4 - 5 | 1240 | 83 | ||
| AK046122.1 | - | 4 - 5 | 1293 | 83 | ||
| AK048634.1 | - | 4 - 5 | 1257 | 83 | ||
| AK133667.1 | - | 4 - 5 | 1272 | 83 | ||
| BC031822.1 | - | 3 - 4 | 708 | 4501004 | 83 | |
| BC049275.1 | - | 3 - 4 | 708 | 4501004 | 83 | |
| X66118.1 | - | 4 - 5 | 1272 | 83 | ||
More Information
Set Membership: |
|

Grik1
Genomic Map