Assay Details
| Gene Symbol: Ppm1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008014.3 | NP_032040.1 | 2 - 3 | 311 | 112 | |
| GenBank mRNA | AK180043.1 | - | 2 - 3 | 99 | 112 | |
| AK183594.1 | - | 2 - 3 | 99 | 112 | ||
| BC009004.1 | - | 2 - 3 | 311 | 3499081 | 112 | |
| U42383.1 | - | 2 - 3 | 197 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1g
Genomic Map