Assay Details
Gene Symbol: Ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008014.3 | NP_032040.1 | 7 - 8 | 1312 | 102 | |
GenBank mRNA | AK187638.1 | - | 1 - 2 | 123 | 102 | |
BC009004.1 | - | 7 - 8 | 1312 | 3499081 | 102 | |
U42383.1 | - | 7 - 8 | 1198 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
