Assay Details
| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008812.2 | NP_032838.2 | 7 - 8 | 936 | 77 | |
| GenBank mRNA | AK148156.1 | - | 7 - 8 | 923 | 77 | |
| AK154814.1 | - | 7 - 8 | 924 | 77 | ||
| AK158382.1 | - | 7 - 8 | 923 | 77 | ||
| AK168685.1 | - | 4 - 5 | 645 | 77 | ||
| AK171275.1 | - | 7 - 8 | 923 | 77 | ||
| BC040350.1 | - | 7 - 8 | 878 | 4192742 | 77 | |
| BC049947.1 | - | 7 - 8 | 909 | 3600978 | 77 | |
| D16580.1 | - | 7 - 8 | 869 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Padi2
Genomic Map