Assay Details
| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008812.2 | NP_032838.2 | 12 - 13 | 1553 | 76 | |
| GenBank mRNA | AK033760.1 | - | 4 - 5 | 526 | 76 | |
| AK148156.1 | - | 12 - 13 | 1540 | 76 | ||
| AK154814.1 | - | 12 - 13 | 1541 | 76 | ||
| AK168685.1 | - | 9 - 10 | 1262 | 76 | ||
| AK171275.1 | - | 12 - 13 | 1540 | 76 | ||
| BC040350.1 | - | 12 - 13 | 1495 | 4192742 | 76 | |
| BC049947.1 | - | 12 - 13 | 1526 | 3600978 | 76 | |
| BC131636.1 | - | 4 - 5 | 434 | 76 | ||
| D16580.1 | - | 12 - 13 | 1486 | 76 | ||
More Information
Set Membership: |
|

Padi2
Genomic Map