Assay Details
| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004701.2 | - | 14 - 15 | 1924 | 60 | |
| AK144750.1 | - | 15 - 16 | 1881 | 60 | ||
| BC003816.1 | - | 15 - 16 | 1921 | 3485483 | 60 | |
| M72394.1 | - | 15 - 16 | 1869 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g4a
Genomic Map