Assay Details
Gene Symbol: Plau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008873.3 | NP_032899.1 | 7 - 8 | 786 | 92 | |
GenBank mRNA | BC120709.1 | - | 6 - 7 | 726 | 40129632 | 92 |
BC120713.1 | - | 6 - 7 | 726 | 40129636 | 92 | |
BC145037.1 | - | 6 - 7 | 749 | 92 | ||
X02389.1 | - | 7 - 8 | 741 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
