Assay Details
Gene Symbol: Plau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008873.3 | NP_032899.1 | 8 - 9 | 943 | 75 | |
GenBank mRNA | BC120709.1 | - | 7 - 8 | 883 | 40129632 | 75 |
BC120713.1 | - | 7 - 8 | 883 | 40129636 | 75 | |
BC145037.1 | - | 7 - 8 | 907 | 75 | ||
X02389.1 | - | 8 - 9 | 898 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
