Assay Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008904.2 | NP_032930.1 | 1 - 2 | 193 | 83 | |
| NR_027710.1 | 1 - 2 | 193 | 83 | |||
| GenBank mRNA | AB061324.1 | - | 1 - 2 | 116 | 83 | |
| AF049330.1 | - | 1 - 2 | 145 | 83 | ||
| AK138668.1 | - | 1 - 2 | 194 | 83 | ||
| AK143753.1 | - | 1 - 2 | 145 | 83 | ||
| BC066868.1 | - | 1 - 2 | 88 | 30094033 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppargc1a
Genomic Map