Assay Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008904.2 | NP_032930.1 | 3 - 4 | 559 | 78 | |
| NR_027710.1 | 3 - 4 | 559 | 78 | |||
| GenBank mRNA | AB061324.1 | - | 3 - 4 | 482 | 78 | |
| AF049330.1 | - | 3 - 4 | 511 | 78 | ||
| AK138668.1 | - | 3 - 4 | 560 | 78 | ||
| AK143753.1 | - | 3 - 4 | 511 | 78 | ||
| BC066868.1 | - | 3 - 4 | 454 | 30094033 | 78 | |
| JX866948.1 | - | 3 - 4 | 447 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppargc1a
Genomic Map