Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100028963.mm.1 |
NM_008911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028963.mm.1 |
NM_008911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028963.mm.1 |
TC0100003536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100028965.mm.1 |
NM_008911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028965.mm.1 |
NM_008911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028965.mm.1 |
TC0100003536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100028962.mm.1 |
NM_008911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028962.mm.1 |
NM_008911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028962.mm.1 |
TC0100003536.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100028967.mm.1 |
NM_008911.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028967.mm.1 |
NM_008911.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028967.mm.1 |
TC0100003536.mm.1 |
MTA TC |