Assay Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008911.2 | NP_032937.1 | 7 - 8 | 1133 | 68 | |
| GenBank mRNA | AK142632.1 | - | 7 - 8 | 1085 | 68 | |
| BC002047.1 | - | 7 - 8 | 1088 | 3484079 | 68 | |
| D45185.1 | - | 7 - 8 | 859 | 68 | ||
| U25114.1 | - | 7 - 8 | 1108 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310949 | D45185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310949 | U25114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310949 | D45185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310949 | U25114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407432 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310949 | D45185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310949 | U25114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310949 | D45185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310949 | U25114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310949 | D45185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310949 | U25114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310949 | D45185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310949 | U25114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827080 | - | - |
| Whole Mouse Genome G4122A | A_51_P310949 | D45185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310949 | U25114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310949 | D45185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310949 | U25114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310949 | BC002047.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131570 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128645 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481666 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481633 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481670 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481630 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00786864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00786863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360092 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360099 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441862_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416618_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360090 | - | - |
| Mouse Genome 430 2.0 Array | 1441862_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448065_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441862_at | - | - |
| Mouse Genome 430 2.0 Array | 1416618_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448065_at | - | - |
| Mouse Genome 430 2.0 Array | 1416618_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19044_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19044_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa17350_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100028948.mm.1 | NM_008911.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028948.mm.1 | NM_008911.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028948.mm.1 | TC0100003536.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416618_PM_at | - | - |
| Mouse Expression Set 430 | 1416618_at | - | - |
| Mouse Expression Set 430 | 1441862_at | - | - |
| Mouse Expression Set 430 | 1441862_PM_at | - | - |
| Murine Genome U74v2 Set | 100429_PM_at | - | - |
| Murine Genome U74v2 Set | 100429_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416618_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416618_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416618_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416618_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441862_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441862_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 820089 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3850102 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850102 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1240672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001240672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706938 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618316 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706938 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618316 | - | - |
| mouse-6 v1.1 (Illumina) | scl15943.8.1_52-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000961.1_62-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1240672 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000961.1_62-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl15943.8.1_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850102 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1240672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2826816 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2706938 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706938 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2826816 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl15943.8.1_52-S | - | - |

Ppox
Genomic Map