Assay Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008911.2 | NP_032937.1 | 10 - 11 | 1422 | 96 | |
| GenBank mRNA | AK142632.1 | - | 10 - 11 | 1374 | 96 | |
| BC002047.1 | - | 10 - 11 | 1377 | 3484079 | 96 | |
| D45185.1 | - | 10 - 11 | 1148 | 96 | ||
| U25114.1 | - | 10 - 11 | 1397 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppox
Genomic Map