Assay Details
Gene Symbol: Ptdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008959.3 | NP_032985.2 | 6 - 7 | 1429 | 84 | |
GenBank mRNA | AF042731.1 | - | 6 - 7 | 844 | 84 | |
AK036990.1 | - | 6 - 7 | 891 | 84 | ||
AK054101.1 | - | 6 - 7 | 868 | 84 | ||
AK085857.1 | - | 6 - 7 | 1430 | 84 | ||
AK086136.1 | - | 6 - 7 | 1088 | 84 | ||
AK137681.1 | - | 6 - 7 | 1099 | 84 | ||
AK172873.1 | - | 6 - 7 | 1406 | 84 | ||
BC003260.1 | - | 6 - 7 | 1066 | 3501141 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
