Assay Details
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012076.1 | - | 2 - 3 | 558 | 90 | |
AK075604.1 | - | 2 - 3 | 557 | 90 | ||
AK135948.1 | - | 2 - 3 | 558 | 90 | ||
BC003793.1 | - | 2 - 3 | 525 | 3601675 | 90 | |
U28016.1 | - | 2 - 3 | 534 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
