Assay Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291268.1 | NP_001278197.1 | 3 - 4 | 910 | 61 | |
| NM_001291269.1 | NP_001278198.1 | 3 - 4 | 619 | 61 | ||
| NM_009330.3 | NP_033356.2 | 3 - 4 | 988 | 61 | ||
| GenBank mRNA | AB008174.1 | - | 3 - 4 | 808 | 61 | |
| AB008175.1 | - | 3 - 4 | 730 | 61 | ||
| AB008176.1 | - | 3 - 4 | 808 | 61 | ||
| AB008177.1 | - | 3 - 4 | 730 | 61 | ||
| AB052659.1 | - | 3 - 4 | 808 | 61 | ||
| AK004837.2 | - | 3 - 4 | 989 | 61 | ||
| AK019258.1 | - | 3 - 4 | 542 | 61 | ||
| BC025189.1 | - | 3 - 4 | 619 | 5044299 | 61 | |
| X55842.1 | - | 3 - 4 | 808 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnf1b
Genomic Map