Assay Details
| Gene Symbol: Tfam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009360.4 | NP_033386.1 | 4 - 5 | 534 | 120 | |
| GenBank mRNA | AK004857.1 | - | 4 - 5 | 532 | 120 | |
| AK017974.1 | - | 4 - 5 | 561 | 121 | ||
| AK050446.1 | - | 4 - 5 | 535 | 120 | ||
| AK161747.1 | - | 4 - 5 | 561 | 120 | ||
| AK163563.1 | - | 4 - 5 | 552 | 120 | ||
| AK167348.1 | - | 4 - 5 | 535 | 120 | ||
| AK167777.1 | - | 4 - 5 | 561 | 120 | ||
| AK169808.1 | - | 4 - 5 | 562 | 120 | ||
| BC001987.1 | - | 4 - 5 | 529 | 3487433 | 120 | |
| BC083084.1 | - | 4 - 5 | 529 | 6514962 | 120 | |
| L07107.1 | - | 3 - 4 | 331 | 120 | ||
| U57939.1 | - | 4 - 5 | 526 | 120 | ||
| U63858.1 | - | 4 - 5 | 368 | 120 | ||
| U63859.1 | - | 4 - 5 | 734 | 120 | ||
| U63860.1 | - | 2 - 3 | 162 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfam
Genomic Map