Assay Details
Gene Symbol: Top3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009410.2 | NP_033436.1 | 14 - 15 | 1981 | 62 | |
GenBank mRNA | AB006074.1 | - | 14 - 15 | 1982 | 62 | |
AK029987.1 | - | 11 - 12 | 1374 | 62 | ||
AK037640.1 | - | 14 - 15 | 1882 | 62 | ||
AK164443.1 | - | 14 - 15 | 1882 | 62 | ||
AK170291.1 | - | 15 - 16 | 1788 | 62 | ||
AK171286.1 | - | 15 - 16 | 1788 | 62 | ||
BC099689.1 | - | 14 - 15 | 1796 | 30731611 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
