Assay Details
| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009466.2 | NP_033492.1 | 1 - 2 | 221 | 68 | |
| GenBank mRNA | AF061017.1 | - | 1 - 2 | 105 | 68 | |
| AK144673.1 | - | 1 - 2 | 257 | 68 | ||
| AK146693.1 | - | 1 - 2 | 105 | 68 | ||
| AK162298.1 | - | 1 - 2 | 109 | 68 | ||
| AK167462.1 | - | 1 - 2 | 101 | 68 | ||
| AK167560.1 | - | 1 - 2 | 201 | 68 | ||
| AK168508.1 | - | 1 - 2 | 108 | 68 | ||
| AK168563.1 | - | 1 - 2 | 105 | 68 | ||
| AK169208.1 | - | 1 - 2 | 236 | 68 | ||
| BC006749.1 | - | 1 - 2 | 221 | 3708709 | 68 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF061017.1,AK144673.1,AK146693.1,AK162298.1,AK167462.1,AK167560.1,AK168508.1,AK169208.1,BC006749.1,NM_009466.2 transcripts.
Set Membership: |
|

Ugdh
Genomic Map