Assay Details
| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009466.2 | NP_033492.1 | 9 - 10 | 1398 | 70 | |
| GenBank mRNA | AF061017.1 | - | 9 - 10 | 1282 | 70 | |
| AK144673.1 | - | 9 - 10 | 1434 | 70 | ||
| AK146693.1 | - | 9 - 10 | 1282 | 70 | ||
| AK149590.1 | - | 3 - 4 | 2678 | 70 | ||
| AK162298.1 | - | 9 - 10 | 1286 | 70 | ||
| AK167462.1 | - | 9 - 10 | 1278 | 70 | ||
| AK167560.1 | - | 9 - 10 | 1378 | 70 | ||
| AK168064.1 | - | 7 - 8 | 995 | 70 | ||
| AK168508.1 | - | 9 - 10 | 1285 | 70 | ||
| AK168563.1 | - | 9 - 10 | 1282 | 70 | ||
| AK169208.1 | - | 9 - 10 | 1413 | 70 | ||
| BC006749.1 | - | 9 - 10 | 1398 | 3708709 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugdh
Genomic Map