Assay Details
| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009466.2 | NP_033492.1 | 10 - 11 | 1481 | 93 | |
| GenBank mRNA | AF061017.1 | - | 10 - 11 | 1365 | 93 | |
| AK144673.1 | - | 10 - 11 | 1517 | 93 | ||
| AK146693.1 | - | 10 - 11 | 1365 | 93 | ||
| AK149590.1 | - | 4 - 5 | 2761 | 93 | ||
| AK162298.1 | - | 10 - 11 | 1369 | 93 | ||
| AK167462.1 | - | 10 - 11 | 1361 | 93 | ||
| AK167560.1 | - | 10 - 11 | 1461 | 93 | ||
| AK168064.1 | - | 8 - 9 | 1078 | 93 | ||
| AK168508.1 | - | 10 - 11 | 1368 | 93 | ||
| AK168563.1 | - | 10 - 11 | 1365 | 94 | ||
| AK169208.1 | - | 10 - 11 | 1496 | 93 | ||
| AK190373.1 | - | 1 - 2 | 96 | 93 | ||
| BC006749.1 | - | 10 - 11 | 1481 | 3708709 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugdh
Genomic Map