Assay Details
| Gene Symbol: Urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009478.3 | NP_033504.2 | 2 - 3 | 156 | 65 | |
| GenBank mRNA | AK002844.1 | - | 2 - 3 | 146 | 65 | |
| AK007470.1 | - | 2 - 3 | 146 | 65 | ||
| AK172426.1 | - | 2 - 3 | 167 | 65 | ||
| BC008109.1 | - | 2 - 3 | 156 | 3481699 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Urod
Genomic Map