Assay Details
| Gene Symbol: Urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009478.3 | NP_033504.2 | 6 - 7 | 661 | 80 | |
| GenBank mRNA | AK002844.1 | - | 6 - 7 | 651 | 80 | |
| AK007470.1 | - | 6 - 7 | 651 | 80 | ||
| AK135090.1 | - | 2 - 3 | 493 | 80 | ||
| AK172426.1 | - | 6 - 7 | 672 | 80 | ||
| AK211828.1 | - | 1 - 2 | 164 | 80 | ||
| BC008109.1 | - | 6 - 7 | 661 | 3481699 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856947 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162692 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162692 | - | - |
| Whole Mouse Genome G4122A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134060 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162692 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798073 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03219573 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03755530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03261655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04037950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03798082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04037944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23231893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515388 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417206_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443849_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515385 | - | - |
| Mouse Genome 430 2.0 Array | 1417206_at | - | - |
| Mouse Genome 430 2.0 Array | 1443849_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1443849_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417206_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22275_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22275_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400026835.mm.1 | TC0400003357.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026835.mm.1 | NM_009478.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026832.mm.1 | TC0400003357.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026832.mm.1 | NM_009478.4 | Refseq |
| Mouse Expression Set 430 | 1417206_PM_at | - | - |
| Mouse Expression Set 430 | 1417206_at | - | - |
| Mouse Expression Set 430 | 1443849_PM_x_at | - | - |
| Mouse Expression Set 430 | 1443849_x_at | - | - |
| Murine Genome U74v2 Set | 94275_PM_at | - | - |
| Murine Genome U74v2 Set | 161328_i_at | - | - |
| Murine Genome U74v2 Set | 161328_PM_i_at | - | - |
| Murine Genome U74v2 Set | 94275_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417206_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417206_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417206_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417206_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443849_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443849_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 672607 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2470494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002470494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2453695 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2453695 | - | - |
| mouse-6 v1.1 (Illumina) | scl23946.8.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2470494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23946.8.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2453695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2453695 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23946.8.1_41-S | - | - |

Urod
Genomic Map