Assay Details
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010811.2 | NP_034941.2 | 12 - 13 | 3064 | 72 | |
GenBank mRNA | AF074925.1 | - | 10 - 11 | 2314 | 72 | |
AK150100.1 | - | 12 - 13 | 2845 | 72 | ||
BC110479.1 | - | 10 - 11 | 2400 | 40048114 | 72 | |
BC110480.1 | - | 10 - 11 | 2462 | 40048115 | 72 | |
U02304.1 | - | 11 - 12 | 2826 | 72 | ||
X75885.1 | - | 10 - 11 | 2468 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
